Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin A Mouse anti-Human, Clone: 9G9, Novus Biologicals™

Mouse Monoclonal Antibody

$116.82 - $343.85

Specifications

Antigen Cyclophilin A
Clone 9G9
Immunogen The specific Immunogen is proprietary information.
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17918220 View Documents Novus Biologicals
NBP17918220UL
20ul Each for $116.82
Add to cart
 
NBP179182 View Documents Novus Biologicals
NBP179182
100ul Each for $343.85
Add to cart
 
Description

Description

Cyclophilin A Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Cyclophilin A
The specific Immunogen is proprietary information.
Primary
Unconjugated
5478
Purified
Western Blot
RUO
9G9
Mouse
Monoclonal
Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin
PBS and 2% Sucrose with 0.09% Sodium Azide
Protein A or G purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit