Learn More
Cyclosporin A Mouse anti-Nonspecies specific, FITC, Clone: CSZ22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1102569F
Catalog No. NB1102569F
Add to cart
Description
Description
Cyclosporin A Monoclonal specifically detects Cyclosporin A in Non-species specific samples. It is validated for ELISA.
Specifications
Specifications
Cyclosporin A
CSZ22
ELISA
Purified
PPIA
Cyclophilin A conjugated to KLH
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This recognises Cyclophilin A in buffer solutions and whole blood extracts with Kd = 10^-8
ELISA
FITC
PBS with 0.05% Sodium Azide
Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin
Mouse
IgG1
0.2 ml
Cancer, Cell Cycle and Replication, Stem Cell Markers
Primary
5478
Nonspecies specific