Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin A Rabbit anti-Human, Clone: 4G3, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA538391

Encompass

Catalog No. PIMA538391


Add to cart

Description

Description

This antibody has been tested in direct-ELISA

Cyclophilin A Recombinant Monoclonal antibody specifically detects Cyclophilin A in Human samples. It is validated for Western Blot
Specifications

Specifications

Cyclophilin A
Recombinant Monoclonal
0.49 mg/mL
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide
P62937
2700098C05; Cphn; CYCA; cyclophilin; Cyclophilin A; Cyclosporin A-binding protein; CyP A; CyP-18; CypA; CyP-A; CYPH; EC 5.2.1.8; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; HGNC:9253; I79_011869; MGC117158; MGC12404; MGC23397; p1B15; p31; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase; PPIase A; rotamase; Rotamase A; SP18; T cell cyclophilin
A synthesized peptide derived from human Cyclophilin A
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
4G3
Unconjugated
Ppia
Liquid, Affinity Purified
Rabbit
IgG
100 μL
-20°C or -80°C if preferred
Primary
5478
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit