Learn More
Cyclophilin-F Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300362AF488
Catalog No. NB065743
Add to cart
Description
Description
Cyclophilin-F Polyclonal specifically detects Cyclophilin-F in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Cyclophilin-F
Alexa Fluor 488
50mM Sodium Borate
Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F)
Produced in rabbits immunized with purified, recombinant Cyclophilin-F (NP_005720.1; Cys30-Ser207).
Affinity purified
RUO
Primary
10105
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human