Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophosphamide, Monohydrate, Antineoplastic, USP, 98%, Spectrum™

Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle

C1244, 6055-19-2, C7H15Cl2N2OP•H2O

Spectrum™ Cyclophosphamide, Monohydrate, Antineoplastic, USP
CAS 6055-19-2
CAS Max % 1
Molecular Formula C7H17Cl2N2O3P
UN Number UN2811
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
IUPAC Name 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
Molecular Weight (g/mol) 279.10
Percent Purity 98%
Packaging Amber Glass Bottle
Quantity 100 g
Show More Show Less