Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopropanecarbonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Density | 0.905 |
Boiling Point | 134°C to 135°C |
Flash Point | 32°C (89°F) |
Odor | Recognizable |
Refractive Index | 1.421 |
Quantity | 250 g |
UN Number | UN3275 |
Beilstein | 635790 |
Solubility Information | Miscible with water. |
Formula Weight | 67.09 |
Show More |
Safety and Handling
Hazard Category | H226-H301+H311+H331-H315-H319-H335 |
Hazard Statement | GHS H Statement H300-H310-H330-H226 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: III |
EINECSNumber | 226-836-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction