Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopropanecarbonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1194822

Quantity 100g

 View more versions of this product

Catalog No. AAA1194822


  Check Availability
Add to cart

Specifications

Description

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Soluble in water (reacts violently).

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
Description & Specifications

Chemical Identifiers

4023-34-1
104.54
UN2920
Cyclopropanecarboxylic acid chloride
C4H5ClO
MFCD00001277
471286
Cyclopropanecarbonyl chloride

Specifications

23°C (73°F)
119° to 120°C
4023-34-1
104.54
UN2920
Cyclopropanecarboxylic acid chloride
98%
Pungent
100g
C4H5ClO
MFCD00001277
471286
Cyclopropanecarbonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit