Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Soluble in water (reacts violently).
Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.
Solubility
Soluble in water (reacts violently).
Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
Specifications
Density | 1.15 |
Boiling Point | 119°C to 120°C |
Flash Point | 23°C (73°F) |
Odor | Pungent |
Refractive Index | 1.452 |
Quantity | 25 g |
UN Number | UN2920 |
Beilstein | 471286 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water (reacts violently). |
Show More |
Safety and Handling
Hazard Category | H226-H301-H314 |
Hazard Statement | GHS H Statement H301-H314-H318-H226 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
EINECSNumber | 223-684-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction