Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanecarboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1489-69-6
C4H6O
70.09
MFCD00012261
JMYVMOUINOAAPA-UHFFFAOYSA-N
cyclopropanecarboxaldehyde,formylcyclopropane,cyclopropylmethanal,cyclopropanecarboxyldehyde,cyclopropanal,cyclopropane aldehyde,cyclopropancarbaldehyd,cyclopropancarbaldehyde,cyclopropancarboxaldehyde,cyclopropanecarboaldehyde
123114
cyclopropanecarbaldehyde
C1CC1C=O
1489-69-6
C4H6O
70.09
MFCD00012261
JMYVMOUINOAAPA-UHFFFAOYSA-N
cyclopropanecarboxaldehyde,formylcyclopropane,cyclopropylmethanal,cyclopropanecarboxyldehyde,cyclopropanal,cyclopropane aldehyde,cyclopropancarbaldehyd,cyclopropancarbaldehyde,cyclopropancarboxaldehyde,cyclopropanecarboaldehyde
123114
cyclopropanecarbaldehyde
C1CC1C=O
Density 0.9380g/mL
Quantity 5g
Refractive Index 1.4285 to 1.4305
Boiling Point 98.0°C to 101.0°C
Packaging Glass bottle
Flash Point 7°C
Specific Gravity 0.938
Assay Percent Range 97.5% min. (GC)
Linear Formula C3H5CHO
Solubility Information Solubility in water: insoluble.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger