Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopropanecarboxamide, 98%

Manufacturer:  Alfa Aesar™ B2187118

Quantity 50g

 View more versions of this product

Catalog No. AAB2187118


  Check Availability
Add to cart

Specifications

Description

Applications
Cyclopropanecarboxamide is used in the preparation of 2-(cyclopropanecarbonyl-amino)-but-2-enoic acid by reacting with 2-oxo-butyric acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases, strong reducing agents, acid chlorides and acid anhydrides.
Description & Specifications

Chemical Identifiers

6228-73-5
85.11
1924346
C4H7NO
MFCD00013729
Cyclopropanecarboxamide

Specifications

122° to 126°C
6228-73-5
85.11
1924346
98%
50g
C4H7NO
MFCD00013729
Cyclopropanecarboxamide

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit