Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopropanecarboxamide, 98%

Click to view available options
Quantity:
2g
10g
50g
250g
6228-73-5
C4H7NO
85.106
MFCD00013729
AIMMVWOEOZMVMS-UHFFFAOYSA-N
cyclopropylcarboxamide,carbamoylcyclopropane,cyclopropyl carboxamide,seed one,cyclopropanecarboxylic acid amide,cyclopropanamide,cyclopropancarboxamide,carbamoyl cyclopropane,cyclopropane carboxamide,pubchem21401
80376
CHEBI:51457
cyclopropanecarboxamide
C1CC1C(=O)N
6228-73-5
C4H7NO
85.106
MFCD00013729
AIMMVWOEOZMVMS-UHFFFAOYSA-N
cyclopropylcarboxamide,carbamoylcyclopropane,cyclopropyl carboxamide,seed one,cyclopropanecarboxylic acid amide,cyclopropanamide,cyclopropancarboxamide,carbamoyl cyclopropane,cyclopropane carboxamide,pubchem21401
80376
CHEBI:51457
cyclopropanecarboxamide
C1CC1C(=O)N
Applications
Cyclopropanecarboxamide is used in the preparation of 2-(cyclopropanecarbonyl-amino)-but-2-enoic acid by reacting with 2-oxo-butyric acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases, strong reducing agents, acid chlorides and acid anhydrides.
Melting Point 122°C to 126°C
Quantity 10g
Beilstein 1924346
Formula Weight 85.11
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxamide