Alfa Aesar™ Cyclopropanesulfonamide, 97%

Manufacturer: Alfa Aesar™ H55507MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55507MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 154350-28-4
    Molecular Formula C3H7NO2S
    Formula Weight 121.15
    MDL Number MFCD08705286
    Chemical Name or Material Cyclopropanesulfonamide

    Specifications

    Formula Weight 121.15
    Molecular Formula C3H7NO2S
    MDL Number MFCD08705286
    CAS 154350-28-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 101° to 106°C
    Quantity 250mg
    Chemical Name or Material Cyclopropanesulfonamide
    Assay 97%

    Description

    Applications
    Cyclopropanesulfonamide is used as fine chemicals to the pharmaceutical, agrochemical, aroma, polymer and electronic industries.

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit