Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanesulfonyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
139631-62-2
C3H5ClO2S
140.59
PFWWSGFPICCWGU-UHFFFAOYSA-N
cyclopropanesulfonylchloride,chlorocyclopropylsulfone,cyclopropanesulphonyl chloride,cyclopropylsulfonyl chloride,cyclopropylsulphonyl chloride,cyclopropane sulfonyl chloride,cyclopropanesulfonic acid chloride,pubchem10869,acmc-209cje,cyclopropylsulfonylchloride
2760919
cyclopropanesulfonyl chloride
C1CC1S(=O)(=O)Cl
139631-62-2
C3H5ClO2S
140.59
PFWWSGFPICCWGU-UHFFFAOYSA-N
cyclopropanesulfonylchloride,chlorocyclopropylsulfone,cyclopropanesulphonyl chloride,cyclopropylsulfonyl chloride,cyclopropylsulphonyl chloride,cyclopropane sulfonyl chloride,cyclopropanesulfonic acid chloride,pubchem10869,acmc-209cje,cyclopropylsulfonylchloride
2760919
cyclopropanesulfonyl chloride
C1CC1S(=O)(=O)Cl
Specific Gravity 1.38
Packaging Glass bottle
Quantity 5g
Flash Point 96°C
Density 1.3800g/mL
Color Colorless to Yellow
Boiling Point 60.0°C (2.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Formula Weight 140.59
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Cyclopropanesulfonyl chloride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger