Alfa Aesar™ Cyclopropyl 2-thienyl ketone, 97%

Manufacturer: Alfa Aesar™ H5526206

Quantity 5g

 View more versions of this product

Catalog No. AAH5526206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6193-47-1
    Formula Weight 152.21
    MDL Number MFCD00005427
    Chemical Name or Material Cyclopropyl 2-thienyl ketone

    Specifications

    Formula Weight 152.21
    Physical Form Liquid
    Refractive Index 1.5840
    MDL Number MFCD00005427
    Density 1.17g/mL
    Quantity 5g
    CAS 6193-47-1
    Chemical Name or Material Cyclopropyl 2-thienyl ketone
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit