Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl Isothiocyanate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
56601-42-4
C4H5NS
99.16
MFCD00040874
JGFBQFKZKSSODQ-UHFFFAOYSA-N
cyclopropyl isothiocyanate,cyclopropane, isothiocyanato,isothiocyanato-cyclopropane,cyclopropylisothiocyanate,acmc-20albt,pubchem22785,cyclopropyl iso-thiocyanate,cyclopropane,isothiocyanato
92463
isothiocyanatocyclopropane
C1CC1N=C=S
56601-42-4
C4H5NS
99.16
MFCD00040874
JGFBQFKZKSSODQ-UHFFFAOYSA-N
cyclopropyl isothiocyanate,cyclopropane, isothiocyanato,isothiocyanato-cyclopropane,cyclopropylisothiocyanate,acmc-20albt,pubchem22785,cyclopropyl iso-thiocyanate,cyclopropane,isothiocyanato
92463
isothiocyanatocyclopropane
C1CC1N=C=S
Specific Gravity 1.07
Quantity 1g
Packaging Glass bottle
Density 1.0700g/mL
Refractive Index 1.5440 to 1.5460
Boiling Point 57.0°C to 59.0°C (15.0mmHg)
Infrared Spectrum Authentic
Color Colorless to Yellow
Assay Percent Range 96% min. (GC)
Linear Formula C3H5NCS
Formula Weight 99.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 260-278-6