Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl(trimethylsilyl)acetylene, +97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
81166-84-9
C8H14Si
138.285
MFCD00671355
IYQSVLJDYLPMCX-UHFFFAOYSA-N
cyclopropyl trimethylsilyl acetylene,cyclopropylethynyl trimethylsilane,2-cyclopropylethynyl trimethylsilane,cyclopropylethynyl trimethyl silane,silane, cyclopropylethynyl trimethyl,cyclopropane, 2-trimethylsilyl ethynyl,silane, cyclopropylethynyl trimethyl-9ci,timtec-bb sbb009013,trimethylsilylethynylcyclopropane,cyclopropylethynyl-trimethylsilane
2758018
2-cyclopropylethynyl(trimethyl)silane
C[Si](C)(C)C#CC1CC1
81166-84-9
C8H14Si
138.285
MFCD00671355
IYQSVLJDYLPMCX-UHFFFAOYSA-N
cyclopropyl trimethylsilyl acetylene,cyclopropylethynyl trimethylsilane,2-cyclopropylethynyl trimethylsilane,cyclopropylethynyl trimethyl silane,silane, cyclopropylethynyl trimethyl,cyclopropane, 2-trimethylsilyl ethynyl,silane, cyclopropylethynyl trimethyl-9ci,timtec-bb sbb009013,trimethylsilylethynylcyclopropane,cyclopropylethynyl-trimethylsilane
2758018
2-cyclopropylethynyl(trimethyl)silane
C[Si](C)(C)C#CC1CC1
Refractive Index 1.4530 to 1.4550
Boiling Point 83.0°C (80.0 mmHg)
Flash Point 25°C
Quantity 1g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Linear Formula C3H5CCSi(CH3)3
Formula Weight 138.28
Physical Form Liquid
Percent Purity ≥97%
Chemical Name or Material Cyclopropyl(trimethylsilyl)acetylene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning