Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyldiphenyl sulfonium tetrafluoroborate, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
33462-81-6
C15H15BF4S
314.149
MFCD00011873
DWEYKSWKFYZEGZ-UHFFFAOYSA-N
cyclopropyldiphenylsulfonium tetrafluoroborate,cyclopropyldiphenylsulphonium tetrafluoroborate,cyclopropyldiphenylsulfoniumtetrafluoroborate,cyclopropyldiphenylsulfanium tetrafluoroborate,acmc-1agq3,cyclopropyldiphenylsulfonium fluoroborate,cyclopropyl diphenyl sulfonium tetrafluoroborate,cyclopropyl diphenyl sulfanium tetrafluoroborate,cyclopropyl diphenyl sulphoniumtetrafluoroborate
2724210
cyclopropyl(diphenyl)sulfanium;tetrafluoroborate
[B-](F)(F)(F)F.C1CC1[S+](C2=CC=CC=C2)C3=CC=CC=C3
33462-81-6
C15H15BF4S
314.149
MFCD00011873
DWEYKSWKFYZEGZ-UHFFFAOYSA-N
cyclopropyldiphenylsulfonium tetrafluoroborate,cyclopropyldiphenylsulphonium tetrafluoroborate,cyclopropyldiphenylsulfoniumtetrafluoroborate,cyclopropyldiphenylsulfanium tetrafluoroborate,acmc-1agq3,cyclopropyldiphenylsulfonium fluoroborate,cyclopropyl diphenyl sulfonium tetrafluoroborate,cyclopropyl diphenyl sulfanium tetrafluoroborate,cyclopropyl diphenyl sulphoniumtetrafluoroborate
2724210
cyclopropyl(diphenyl)sulfanium;tetrafluoroborate
[B-](F)(F)(F)F.C1CC1[S+](C2=CC=CC=C2)C3=CC=CC=C3
Color White
Melting Point 137°C to 141°C
Infrared Spectrum Authentic
Quantity 250mg
Assay Percent Range 98%
Linear Formula C3H5S(C6H5)2BF4
Formula Weight 314.14
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Cyclopropyldiphenyl sulfonium tetrafluoroborate, 98%