Alfa Aesar™ Cyclopropylmagnesium bromide, 0.5M-0.7M in THF

Manufacturer: Alfa Aesar™ H26273AC

Quantity 25mL

 View more versions of this product

Catalog No. AAH26273AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23719-80-4
    Molecular Formula C3H5BrMg
    Formula Weight 145.28
    MDL Number MFCD00672094
    UN Number UN2924
    Chemical Name or Material Cyclopropylmagnesium bromide

    Specifications

    Formula Weight 145.28
    MDL Number MFCD00672094
    UN Number UN2924
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 0.5 to 0.7M in THF
    Quantity 25mL
    Molecular Formula C3H5BrMg
    CAS 23719-80-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Flash Point −17°C (1°F)
    Chemical Name or Material Cyclopropylmagnesium bromide

    Description

    Applications
    It often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.

    Solubility
    Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.

    Notes
    Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.

    GHS H Statement
    H225-H314-H351-H335
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit