Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.
Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.
It often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.
Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.
Specifications
Density | 0.968 |
Flash Point | −17°C (1°F) |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive; May form a precipitate |
Solubility Information | Soluble in water (17mg/ml),(25°C). Soluble in organic solvents. |
Formula Weight | 145.28 |
Concentration or Composition (by Analyte or Components) | 0.5 to 0.7M in THF |
Chemical Name or Material | Cyclopropylmagnesium bromide |
Safety and Handling
Hazard Category | H225-H260-H302-H314-H335-H336-H351 |
Hazard Statement | GHS H Statement H225-H314-H351-H335 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.3; Packaging Group: I |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction