Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopropylmagnesium bromide, 1.0M in 2-MeTHF

Click to view available options
Quantity:
25mL
100mL
23719-80-4
C3H5BrMg
145.282
MFCD00672094
VFZXMEQGIIWBFJ-UHFFFAOYSA-M
cyclopropylmagnesium bromide,cyclopropylmagnesiumbromide,bromo cyclopropyl magnesium,magnesium, bromocyclopropyl,cyclopropyl magnesium bromide,cprmgbr,cyclopropyl magnesiumbromide,magnesium,bromocyclopropyl,bromo-cyclopropyl magnesium,cyclopropylmagnesiunn bromide
11116189
magnesium;cyclopropane;bromide
C1C[CH-]1.[Mg+2].[Br-]
23719-80-4
C3H5BrMg
145.282
MFCD00672094
VFZXMEQGIIWBFJ-UHFFFAOYSA-M
cyclopropylmagnesium bromide,cyclopropylmagnesiumbromide,bromo cyclopropyl magnesium,magnesium, bromocyclopropyl,cyclopropyl magnesium bromide,cprmgbr,cyclopropyl magnesiumbromide,magnesium,bromocyclopropyl,bromo-cyclopropyl magnesium,cyclopropylmagnesiunn bromide
11116189
magnesium;cyclopropane;bromide
C1C[CH-]1.[Mg+2].[Br-]
Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Color Yellow to Green
Density 1.022g/mL
Quantity 100mL
Sensitivity Air & Moisture Sensitive
UN Number UN2924
Concentration or Composition (by Analyte or Components) 1.0M in 2-MeTHF
Formula Weight 145.28
Physical Form Turbid Liquid
Chemical Name or Material Cyclopropylmagnesium bromide

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

storageNote1 : 2° to 8°C