Learn More
CYCS Mouse anti-Horse, Human, Clone: 7H8.2C12, Abnova™
Mouse monoclonal antibody raised against horse CYCS.
Manufacturer:  Abnova Corporation MAB12912
Catalog No. 89-934-030
Add to cart
Description
Description
Specifications
Specifications
CYCS
7H8.2C12
Mouse monoclonal antibody raised against horse CYCS.
In PBS, pH 7.2 (50% glycerol, 1% BSA, 0.09% sodium azide).
CYCS
Horse CYCS.
Protein G purification
RUO
Primary
100053958
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG2a
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Equine, Human