Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYCS Mouse anti-Human, Clone: CYCS/1010, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14396
Description
Mouse monoclonal antibody raised against full length recombinant human CYCS.
- High specificity
- High reactivity
Specifications
CYCS | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be d | |
P99999 | |
CYCS | |
Recombinant protein corresponding to full length human CYCS. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
CYCS/1010 | |
Mouse monoclonal antibody raised against full length recombinant human CYCS. | |
In 1 mM PBS. | |
CYC/HCS | |
Mouse | |
Protein A/G purification | |
RUO | |
54205 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction