Learn More
CYCS Mouse anti-Human, Pigeon, Clone: 7H8.2C12, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CYCS.
Manufacturer:  Abnova Corporation MAB13398
Catalog No. 89-934-029
Add to cart
Description
Description
This gene encodes cytochrome c, a component of the electron transport chain in mitochondria. The heme group of cytochrome c accepts electrons from the b-c1 complex and transfers electrons to the cytochrome oxidase complex. Cytochrome c is also involved in initiation of apoptosis. Upon release of cytochrome c to the cytoplasm, the protein binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Many cytochrome c pseudogenes exist, scattered throughout the human genome. [provided by RefSeq]
Specifications
Specifications
CYCS
7H8.2C12
Mouse monoclonal antibody raised against synthetic peptide of CYCS.
In 1mM PBS.
CYC/HCS
Mouse
IgG2b, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Pigeon
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be d
Liquid
CYCS
A synthetic peptide corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYCS.
Protein A/G purification
RUO
Primary
54205