Learn More
Cytiva CyDye™ DIGE Fluor Preparative Gel Labeling for Scarce Samples
For pre-labeling protein samples prior to 2D electrophoresis for difference analysis that enables multiplexing within the same 2D gel.
Manufacturer:  Cytiva 28936683
Catalog No. 45-005-932
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 400nmol CyDye DIGE fluor Cy3 saturation dye for preparative labeling
Description
Description
Cy3 saturation dye is size- and charge-matched fluorescent dye specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.
  • Bright and highly sensitive dyes
    Order Info
    Dimethylformamide (DMF) has to be ordered separately.
Specifications
Specifications
CyDye DIGE Fluor Preparative Gel Labeling for Scarce Samples
400nmol CyDye DIGE fluor Cy3 saturation dye for preparative labeling
Red
Pre-labeling protein samples prior to 2D electrophoresis for difference analysis that enables multiplexing within the same 2D gel
400 nmol Vial