Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYFIP1 Rabbit anti-Human, Mouse, Rat, Clone: S08-9B4, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | CYFIP1 |
---|---|
Clone | S08-9B4 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CYFIP1 Monoclonal antibody specifically detects CYFIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
CYFIP1 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
cytoplasmic FMR1 interacting protein 1, cytoplasmic FMR1-interacting protein 1, FLJ45151, KIAA0068cytoplasmic FMRP interacting protein 1, P140SRA-1, selective hybridizing clone, SHYC, Specifically Rac1-associated protein 1, Sra-1, SRA1 | |
A synthetic peptide of human CYFIP1 (Uniprot#: Q7L576) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S08-9B4 | |
Monoclonal | |
Purified | |
RUO | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
23191 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title