Learn More
Andwin ScientificSupplier Diversity Partner CYLINDER GRAD PP 250ML 12/CS DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner 65000008
Polypropylene Graduated Cylinders 250mL cylinders feature a single scale with raised numeral graduations in 2mL increments. Non-wetting polypropylene surface eliminates concave menisci. Flared top rim. Large spout. Hexagonal base. Chemically resistant
Catalog No. NC0092624