Learn More
Brady™ Cylinder Status Tags: FULL CYLINDER READY FOR USE
Identify facility equipment to your exact specifications. Brady™ Cylinder Status Tags are durable two-sided tags that are an economical and convenient way to ID equipment.
Manufacturer:  Brady™ 17926
Catalog No. 19-113-106
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
3 in. x 5.75 in.
Not laminated
Rectangle
Polyester
White on green
(10) nylon fasteners
10/Pack
-40°F to 212°F
FULL CYLINDER READY FOR USE