Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CYM 5520

Selective S1P2 allosteric agonist

Specifications

Chemical Name or Material 1-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethyl]-1,6-dihydro-6-oxo-3-pyridinecarbonitrile
Molecular Formula C21H19N3O2
CAS 1449747-00-5
Product Type Target: Sphingosine-1-phosphate Receptor Agonists
Formula Weight 345.39
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
54-181-0 View Documents Tocris Bioscience™
5418/10
10mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Selective S1P2 allosteric agonist (EC50 = 0.48 μM). Selective for S1P2 receptors over a panel of 29 other receptors and transporters.
Specifications

Specifications

1-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethyl]-1,6-dihydro-6-oxo-3-pyridinecarbonitrile
1449747-00-5
345.39
C21H19N3O2
Target: Sphingosine-1-phosphate Receptor Agonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit