Learn More
iQ Biosciences Cynomolgus Monkey PBMCs, Monocytes Depleted, 10M cells/vial
Supplier:  iQ Biosciences IQBMNPB102MD
Encompass_Preferred
High-quality, high-viability (> 90%) cynomolgus (cyno) monkey (or macaque) peripheral blood mononuclear cells (PBMCs), with monocytes depleted, sourced responsibly from a vetted third party facility. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor
Catalog No. NC2855588
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.