Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus DcR3/TNFRSF6B Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 9297DC100
This item is not returnable.
View return policy
Description
The Recombinant Cynomolgus DcR3/TNFRSF6B Fc Chimera Protein is derived from CHO. The Recombinant Cynomolgus DcR3/TNFRSF6B Fc Chimera Protein has been validated for the following applications: Bioactivity.Order Info
The product is shipped at ambient temperature.
Specifications
Q9R0Q8 | |
Lyophilized from a 0.2μm filtered solution in PBS | |
56 to 68 kDa, reducing conditions | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.10 EU per 1μg of protein by LAL method | |
Recombinant protein | |
Cynomolgus Monkey | |
No |
Bioactivity | |
26253 | |
DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy | |
Chinese Hamster Ovary cell line,CHO-derived cynomolgus monkey DcR3/TNFRSF6B protein, (N-terminus) Cynomolgus Monkey DcR3/TNFRSF6B (Ala30-His300) Accession &Num; EHH65162 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
RUO | |
8 to 48ng/mL (ED 50) | |
Unconjugated | |
Recombinant | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction