Learn More
R&D Systems™ Recombinant Cynomolgus Monkey B7-1/CD80 His-tag Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9244B1100
Catalog No. 92-44B-1100
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Cynomolgus Monkey B7-1/CD80 Protein is derived from HEK293. The Recombinant Cynomolgus Monkey B7-1/CD80 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
XP_005548122
151888
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
100μg
RUO
0.1 to 0.5μg/mL (ED 50)
Unconjugated
Yes
Bioactivity
15 kDa, reducing conditions
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of protein by LAL method
Recombinant protein
Cynomolgus Monkey
No