Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey B7-H2 His tagged Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 9327B7100
Includes: For Research Use Only
Description
Measured by its ability to co-stimulate IL-4 secretion by D10.G4.1 mouse helper T cells in the presence of anti-CD3. The ED50 for this effect is 0.15-0.9 μg/mLOrder Info
Ships at ambient temperature.
Specifications
XP005548618 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 54 - 70 kDa, reducing conditions; M.W. (theoretical): 27 kDa | |
Human embryonic kidney cell, HEK293-derived cynomolgus monkey B7-H2 protein Asp19-Thr256, with a C-terminal 6-His tag | |
RUO | |
0.15 to 0.9μg/mL | |
Unconjugated | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
23308 | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Recombinant protein | |
Cynomolgus Monkey | |
Reconstitute at 200 μg/mL in PBS. | |
B7-H2/ICOSLG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction