Learn More
R&D Systems™ Cynomolgus Monkey TIM-4 Fc Chimera Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9384TM050
Catalog No. 93-84T-M050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Cynomolgus Monkey TIM-4 Fc Chimera Protein is derived from HEK293. The Recombinant Cynomolgus Monkey TIM-4 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
XP_005558436
Lyophilized from a 0.2μm filtered solution in PBS
57 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of the protein by the LAL method.
0.2 to 1.2μg/mL
Unlabeled
Yes
Bioactivity
91937
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
HEK293
RUO
FLJ27515, SMUCKLER
Recombinant protein
Cynomolgus Monkey
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.