Learn More
Acro Biosystems Recombinant Protein;Cynomolgus / Rhesus macaque TIGIT Protein (HPLC-verified);HEK293;50UG;TIT,TIGIT,VSIG9,VSTM3,VSIG-9,VSTM-3 DFS Item
Manufacturer:  Acro Biosystems TITC522350UG
Recombinant Protein;Cynomolgus / Rhesus macaque TIGIT Protein (HPLC-verified);50UG;Cynomolgus / Rhesus macaque TIGIT (HPLC-verified) (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # XP_015300911.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque TIGIT (HPLC-verified) (Cat. No. TIT-C5223) at 5 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 0.078-2.5 μg/mL (QC tested).
Catalog No. 50-113-8386