Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP11B1 Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP168883
Description
CYP11B1 Polyclonal specifically detects CYP11B1 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CYP11B1 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P15538 | |
CYP11B1 | |
Synthetic peptides corresponding to CYP11B1 (cytochrome P450, family 11, subfamily B, polypeptide 1) The peptide sequence was selected from the C terminal of CYP11B1. Peptide sequence ARNPNVQQALRQESLAAAASISEHPQKATTELPLLRAALKETLRLYPVGL. | |
Affinity Purified | |
RUO | |
Primary | |
Pig: 79%; Sheep: 79%. | |
Human, Mouse, Porcine, Monkey, Sheep | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CPN1, CYPXIB1, cytochrome P450 11B1, mitochondrial, cytochrome p450 XIB1, cytochrome P450, family 11, subfamily B, polypeptide 1, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1, Cytochrome P-450c11, Cytochrome P450C11, EC 1.14.15, EC 1.14.15.4, FHICYP11B, FLJ36771, P450C11DKFZp686B05283, S11BH, Steroid 11-beta-hydroxylase, steroid 11-beta-monooxygenase | |
Rabbit | |
55 kDa | |
100 μL | |
Lipid and Metabolism | |
1584 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title