Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP11B1 Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $436.00

Specifications

Antigen CYP11B1
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16888320
View Documents
Novus Biologicals
NBP16888320UL
20 μL
Each for $152.22
Add to cart
 
NBP168883
View Documents
Novus Biologicals
NBP168883
100 μL
Each for $436.00
Add to cart
 
Description

Description

CYP11B1 Polyclonal specifically detects CYP11B1 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CYP11B1
Unconjugated
RUO
PBS and 2% Sucrose with 0.09% Sodium Azide
CPN1, CYPXIB1, cytochrome P450 11B1, mitochondrial, cytochrome p450 XIB1, cytochrome P450, family 11, subfamily B, polypeptide 1, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1, Cytochrome P-450c11, Cytochrome P450C11, EC 1.14.15, EC 1.14.15.4, FHICYP11B, FLJ36771, P450C11DKFZp686B05283, S11BH, Steroid 11-beta-hydroxylase, steroid 11-beta-monooxygenase
CYP11B1
IgG
Affinity Purified
55 kDa
Polyclonal
Rabbit
Lipid and Metabolism
P15538
1584
Synthetic peptides corresponding to CYP11B1 (cytochrome P450, family 11, subfamily B, polypeptide 1) The peptide sequence was selected from the C terminal of CYP11B1. Peptide sequence ARNPNVQQALRQESLAAAASISEHPQKATTELPLLRAALKETLRLYPVGL.
Primary
Store at -20C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit