Learn More
Aviva Systems Biology Corp CYP20A1 Antibody - N-terminal region (ARP57758_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP20A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYP20A1. Alternate names: CYP-M, MGC22229ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62146