Learn More
Aviva Systems Biology Corp CYP27C1 Antibody - middle region (ARP54373_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYP27C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYP27C1. Alternate names: FLJ16008ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-014-61191