Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2A13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549618

Catalog No. PA549618

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Exhibits a coumarin 7-hydroxylase activity. Active in the metabolic activation of hexamethylphosphoramide, N,N- dimethylaniline, 2 ft.-methoxyacetophenone, N- nitrosomethylphenylamine, and the tobacco-specific carcinogen, 4- (methylnitrosamino)-1-(3-pyridyl)-1-butanone. Possesses phenacetin O-deethylation activity.
Specifications

Specifications

CYP2A13
Polyclonal
PBS with 0.09% sodium azide
Q16696
Cytochrome P450 2A13, CYPIIA13, CYP2A13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1553
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CYP2A13
Liquid
CYP2A13
KLH conjugated synthetic peptide between 446-477 amino acids from the C-terminal region of human CYP2A13.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit