Learn More
BioVision IncSupplier Diversity Partner Cytochrome P450 2C19 (CYP2C19) Activity Assay Kit (Fluorometric): Quantitative 96-well plate format assay of Cytochrome P450 2C19 (CYP2C19) activity in tissues, cells, microsomes or purified enzyme. Simple & Sensitive DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K848100
Cytochrome P450 2C19 (CYP2C19) Activity Assay Kit (Fluorometric): Cytochrome P450 2C19 (CYP2C19, EC 1.14.14.1) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. BioVision's CYP2C19 Activity Assay Kit enables rapid measurement of native or recombinant CYP2C19 activity in biological samples such as liver microsomes. The assay utilizes a non-fluorescent CYP2C19 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 406/468 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. CYP2C19 specific activity is calculated by running parallel reactions in the presence and absence of the selective inhibitor and subtracting any residual activity detected with the inhibitor present. The kit contains a complete set of reagents sufficient for performing 50 sets of paired reactions (in the presence and absence of inhibitor).
Catalog No. 50-111-9960