Learn More
CYP2E1 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for CYP2E1
Manufacturer: Abnova Corporation H00001571T01
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke.
- Cytochrome P450, family 2, subfamily E, polypeptide 1
- Transfected Cell Line: 293T
- Plasmid: pCMV-CYP2E1 full-length
- Host: Human
- Theoretical MW: 56.8kDa
- Storage Buffer: 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Specifications
293T | |
NM_000773.3 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CYP2E1 antibody (H00001571-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Human |
CYP2E1 293T Cell Transient Overexpression Lysate(Denatured) | |
56.8 | |
pCMV-CYP2E1 full-length | |
100 uL | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Western Blot |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.