Learn More
CYP2E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549923
Catalog No. PA549923
Add to cart
Description
Description
The antibody detects endogenous levels of total CYP2E1 protein.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
Specifications
Specifications
CYP2E1
Polyclonal
Unconjugated
CYP2E1
Liquid
CYP2E1
Synthesized peptide derived from C-terminal of human Cytochrome P450 2E1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05182, P05181, Q05421
CP2E1, CYP2E, CYPIIE1, Cytochrome P450 2E1, P450-J
Rabbit
IgG
100 μL
-20°C
Primary
13106, 1571, 25086