Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyp2f2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570471
Description
This target displays homology in the following species: Cow: 86%; Dog: 77%; Goat: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 79%; Rat: 83%.
Cyp2f2 is a protein coding gene involved in the regio- and stereoselective transformation of naphthalene to trans-1R-hydroxy-2R-glutathionyl-1,2-dihydronaphthalene in the presence of glutathione and glutathione S-transferases. It specifically catalyzes the production of a very reactive and potentially toxic intermediate, the 2R,2S arene oxide, that is associated with necrosis of the unciliated bronchiolar epithelial cells or Clara cells in lung.
Specifications
Cyp2f2 | |
Polyclonal | |
Unconjugated | |
Cyp2f2 | |
Cyp2f; Cyp2f1; Cyp2f2; Cyp2f-2; Cyp2f4; CYP4502F4; CYPIIF2; cytochrome P450 2F2; cytochrome P450, 2f2; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, family 2, subfamily f, polypeptide 4; cytochrome P450, subfamily 2F, polypeptide 1; Cytochrome P450, subfamily IIF, polypeptide 1; cytochrome P450-NAH-2; Naphthalene dehydrogenase; naphthalene hydroxylase; P450-NAH-2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13107, 54246 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O35293, P33267 | |
Cyp2f2, Cyp2f4 | |
synthetic peptide directed towards the middle region of human Cyp2f2. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction