Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2J2 Mouse anti-Human, Clone: OTI5C9, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503482S
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is thought to be the predomit enzyme responsible for epoxidation of endogenous arachidonic acid in cardiac tissue.Specifications
CYP2J2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P51589 | |
CPJ2, CYPIIJ2 | |
Mouse | |
IgG1 | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
1573 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5C9 | |
Unconjugated | |
CYP2J2 | |
Liquid | |
CYP2J2 | |
Full length human recombit protein of human CYP2J2 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title