Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2W1 Mouse anti-Human, Alexa Fluor 700, Clone: V32-P5E5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250205AF700
Description
CYP2W1 Monoclonal specifically detects CYP2W1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP2W1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP2W1 | |
Ovalbumin-conjugated synthetic peptide - RYRLLPPPGV (C-terminal sequence) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
V32-P5E5 | |
Western Blot, ELISA, Immunohistochemistry | |
CYPIIW1, cytochrome P450 2W1, cytochrome P450, family 2, subfamily W, polypeptide 1, EC 1.14.14.-, EC 1.14.14.1, EC 1.14.14.5, FLJ20359, MGC34287 | |
Mouse | |
Protein A purified | |
RUO | |
54905 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction