Learn More
CYP2W1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589417
Description
Immunogen sequence: EPVADKILQE LKCLSGQLDG YRGRPFPLAL LGWAPSNITF ALLFGRRFDY RDPVFVSLLG LIDEVMVLLG SPGLQLFNVY PWLGALLQLH RPVLRKIEEV RAILRTLLEA RRPHVCPGDP VCSYVDALIQ QGQGDDPEGL FAEANAVACT LDMVMAGTET TSATLQWAAL LMGRHPDVQG RVQEELDRVL GPGRTPRLED QQALPYTSAV LHEVQRFITL LPHVPRCTAA DTQLGGFLLP KGTPVIPLLT SVLLDETQWQ TPGQFNPGHF LDANGHFVKR EAFLPFSAGR RVCVGERLAR TELFLLFAGL LQRYRLLPPP GVSPASLDTT PARAFTMRPR AQALCAVPRP; Positive Samples: Mouse small intestine; Cellular Location: Cell membrane, Endoplasmic reticulum lumen
CYP2W1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.Specifications
CYP2W1 | |
Polyclonal | |
Unconjugated | |
Cyp2w1 | |
Cyp2w1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 141-490 of human CYP2W1 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
1.81 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9Q816 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
545817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.