Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP3C1 Antibody (V12P6F5*E3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250210H
Description
CYP3C1 Monoclonal specifically detects CYP3C1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP3C1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V12P6F5*E3 | |
Western Blot, ELISA, Immunohistochemistry | |
CYP3A, cyp3c1, Cytochrome P450, family 3, subfamily c, polypeptide 1, wu:fa96d12, wu:fc26b04, zgc:56626 | |
Ovalbumin-conjugated synthetic peptide KLIPRSHKEK (C-terminal sequence) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction