Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP4F12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100893
Description
Antibody detects endogenous levels of total Cytochrome P450 4F12.
CYP4F12 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein likely localizes to the endoplasmic reticulum.
Specifications
CYP4F12 | |
Polyclonal | |
Unconjugated | |
CYP4F12 | |
CYP4F12; CYPIVF12; Cytochrome P450 4F12; cytochrome P450 family 4 subfamily F member 12; cytochrome P450, family 4, subfamily F, polypeptide 12; cytochrome P450, subfamily IVF, polypeptide 12; F22329_1; UNQ568/PRO1129 | |
Rabbit | |
Affinity chromatography | |
RUO | |
66002 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HCS2 | |
CYP4F12 | |
A synthesized peptide derived from human CYP4F12(Accession Q9HCS2), corresponding to amino acid residues C50-V100. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction