Learn More
CYP4Z1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201421AP
Catalog No. 50-173-2728
Add to cart
Description
Description
This antibody is specific to CYP4Z1.
The cytochrome P450 (P450) superfamily of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as nicotinamide adenine dinucleotide phosphate (NADPH) and P450 reductase. There are over 200 known genes which encode P450s. Epoxygenases are those P450s which metabolize AA to epoxyeicosatrienoic acid (EETs) and omega-hydroxylases are those P450s which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). As well as fatty acid metabolism, P450s also metabolize many drugs and toxins. Cytochrome P450 3A4 is abundantly expressed in liver and small intestine and is inducible by barbiturates, glucocorticoids and rifampicin.
Specifications
Specifications
CYP4Z1
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86W10
CYP4A20, CYP4Z1, CYPIVZ1, Cytochrome P450 4Z1
Rabbit
IgG
150 μL
-20°C
Primary
199974
Western Blot
Unconjugated
CYP4Z1
Liquid
CYP4Z1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human