Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP7B1 Antibody (M17-P3F2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250217AF405
Description
CYP7B1 Monoclonal specifically detects CYP7B1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CYP7B1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP7B1 | |
Ovalbumin-conjugated synthetic peptide IQYPDSDVL (C-terminal sequence) | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
9420 | |
Store at 4C in the dark. | |
IgG1 λ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
M17-P3F2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CBAS3, CP7B, Cytochrome P450 7B1, cytochrome P450, family 7, subfamily B, polypeptide 1, cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1,25-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.100, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase, spastic paraplegia 5A (autosomal recessive), SPG5A | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction